Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drosha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130149.1 | NP_001123621.1 | 16 | 2490 | ||
NM_026799.3 | NP_081075.3 | 16 | 2486 | |||
GenBank mRNA | AF533013.1 | - | 12 | 1154 | ||
AK077549.1 | - | 12 | 1294 | |||
AK144147.1 | - | 16 | 2445 | |||
AK148640.1 | - | 16 | 2409 | |||
BC040801.1 | - | 1 | 109 | |||
BC050057.1 | - | 2 | 208 | |||
BC055696.1 | - | 5 | 431 | |||
BC057687.1 | - | 9 | 918 | |||
BC060265.1 | - | 13 | 1785 | |||
BC088999.1 | - | 16 | 2424 | 6417566 |