Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK089402.1 | - | 4 | 349 | ||
AK154742.1 | - | 4 | 349 | |||
AK155323.1 | - | 4 | 333 | |||
AK171362.1 | - | 4 | 346 | |||
BC061149.1 | - | 4 | 332 | 30305571 | ||
U44795.1 | - | 4 | 338 |