Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080848.2 | NP_543124.1 | 10 | 864 | ||
GenBank mRNA | AF416510.1 | - | 10 | 869 | ||
AK075937.1 | - | 10 | 807 | |||
AK144091.1 | - | 10 | 808 | |||
BC008547.1 | - | 5 | 244 | |||
BC016103.1 | - | 10 | 799 | 4919781 | ||
BC025801.1 | - | 10 | 841 | 5132525 |