Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fancc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042673.2 | NP_001036138.1 | 14 | 1795 | ||
NM_001282942.1 | NP_001269871.1 | 13 | 1598 | |||
NM_007985.3 | NP_032011.2 | 14 | 1795 | |||
GenBank mRNA | AK035488.1 | - | 14 | 1549 | ||
AK042226.1 | - | 14 | 1765 | |||
AK082717.1 | - | 11 | 1316 | |||
AK147025.1 | - | 14 | 1775 | |||
BC038609.1 | - | 13 | 1580 | 5375091 | ||
L08266.1 | - | 14 | 1776 |