Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dclre1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025312.1 | NP_001020483.1 | 3 | 1233 | ||
NM_133865.2 | NP_598626.2 | 4 | 1399 | |||
GenBank mRNA | AK049115.1 | - | 4 | 1394 | ||
AK051104.1 | - | 4 | 1354 | |||
AK084347.1 | - | 3 | 1228 | |||
AK134324.1 | - | 4 | 1159 | |||
AK137495.1 | - | 4 | 956 | |||
AK138568.1 | - | 4 | 1457 | |||
AK154124.1 | - | 4 | 1301 | |||
BC011094.1 | - | 3 | 858 | 3493038 | ||
BC067017.1 | - | 3 | 1142 | 6827667 | ||
BC125277.1 | - | 3 | 854 | |||
BC144725.1 | - | 3 | 844 | |||
CA317555.1 | - | 1 | 118 |