Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcgr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077189.1 | NP_001070657.1 | 1 | 107 | ||
NM_010187.2 | NP_034317.1 | 1 | 107 | |||
GenBank mRNA | AK080885.1 | - | 1 | 110 | ||
AK089266.1 | - | 1 | 108 | |||
AK150613.1 | - | 1 | 108 | |||
AK155329.1 | - | 1 | 99 | |||
AK170035.1 | - | 1 | 97 | |||
AK171254.1 | - | 1 | 99 | |||
AK171316.1 | - | 1 | 90 | |||
M14216.1 | - | 1 | 65 | |||
M16367.1 | - | 3 | 348 | |||
M17515.1 | - | 1 | 61 | |||
U51629.1 | - | 1 | 15 |