Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008006.2 | NP_032032.1 | 3 | 580 | ||
GenBank mRNA | AF065903.1 | - | 3 | 383 | ||
AF065904.1 | - | 3 | 383 | |||
AF065905.1 | - | 3 | 383 | |||
AY027548.1 | - | 3 | 298 | |||
AY027549.1 | - | 3 | 403 | |||
AY027550.1 | - | 1 | 175 | |||
AY027551.1 | - | 3 | 380 | |||
AY027557.1 | - | 3 | 453 | |||
AY027558.1 | - | 3 | 444 | |||
AY027559.1 | - | 3 | 307 | |||
BY730691.1 | - | 3 | 558 | |||
M30644.1 | - | 3 | 383 |