Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010202.5 | NP_034332.2 | 3 | 572 | ||
GenBank mRNA | AK082814.1 | - | 3 | 573 | ||
BC104312.1 | - | 3 | 453 | 40044462 | ||
BC104313.1 | - | 3 | 453 | 40044466 | ||
M30642.1 | - | 3 | 453 |