Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF047699.1 | - | 3 | 430 | ||
AF055573.1 | - | 296 | ||||
BC012662.1 | - | 1 | 155 | 4036815 | ||
CJ053929.1 | - | 189 | ||||
CJ056464.1 | - | 4 | 318 | |||
CJ058331.1 | - | 4 | 316 |