Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF047699.1 | - | 4 | 486 | ||
AF055573.1 | - | 352 | ||||
BC012662.1 | - | 2 | 211 | 4036815 | ||
CJ053929.1 | - | 245 | ||||
CJ056464.1 | - | 5 | 374 | |||
CJ058331.1 | - | 5 | 372 | |||
CO040033.1 | - | 5 | 623 |