Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077361.1 | NP_001070829.1 | 3 | 381 | ||
NM_001077362.2 | NP_001070830.1 | 2 | 224 | |||
NM_001287800.1 | NP_001274729.1 | 2 | 168 | |||
NM_010211.3 | NP_034341.2 | 3 | 277 | |||
GenBank mRNA | AF114380.1 | - | 1 | 138 | ||
AK128904.1 | - | 3 | 251 | |||
AK158966.1 | - | 3 | 309 | |||
BC029024.1 | - | 2 | 169 | 5012394 | ||
BC031120.1 | - | 1 | 118 | 4989977 | ||
BC059009.1 | - | 1 | 103 | |||
U41739.1 | - | 2 | 168 | |||
U77039.1 | - | 3 | 221 |