Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077361.1 | NP_001070829.1 | 5, 6 | 807 | ||
NM_001077362.2 | NP_001070830.1 | 4, 5 | 650 | |||
NM_001287800.1 | NP_001274729.1 | 4, 5 | 594 | |||
NM_010211.3 | NP_034341.2 | 5, 6 | 703 | |||
GenBank mRNA | AF114380.1 | - | 3, 4 | 564 | ||
AK128904.1 | - | 5, 6 | 677 | |||
AK158966.1 | - | 5, 6 | 735 | |||
BC029024.1 | - | 4, 5 | 595 | 5012394 | ||
BC031120.1 | - | 3, 4 | 544 | 4989977 | ||
BC059009.1 | - | 2, 3 | 1087 | |||
U41739.1 | - | 4, 5 | 594 | |||
U77039.1 | - | 5, 6 | 647 |