Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010217.2 | NP_034347.2 | 3 | 743 | ||
GenBank mRNA | BC006783.1 | - | 3 | 664 | 3589136 | |
BQ917383.1 | - | 3 | 682 | |||
CJ055589.1 | - | 1 | 122 | |||
M70642.1 | - | 3 | 596 | |||
M80263.1 | - | 3 | 662 |