Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_180974.4 | NP_851305.2 | 2 | 461 | ||
GenBank mRNA | AK030735.1 | - | 2 | 416 | ||
AK031639.1 | - | 2 | 399 | |||
AK154401.1 | - | 2 | 453 | |||
BC138824.1 | - | 2 | 348 | |||
BC138825.1 | - | 2 | 348 | |||
CN698353.1 | - | 2 | 392 |