Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flt3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013520.3 | NP_038548.3 | 6 | 547 | ||
GenBank mRNA | BC019801.1 | - | 6 | 532 | 5132319 | |
CK622267.1 | - | 6 | 517 | |||
EU274583.1 | - | 5 | 459 | |||
L23636.1 | - | 5 | 490 | |||
S76459.1 | - | 5 | 465 | |||
U04807.1 | - | 5 | 714 |