Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Folr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252552.1 | NP_001239481.1 | 7 | 1221 | ||
NM_001252553.1 | NP_001239482.1 | 7 | 1147 | |||
NM_001252554.1 | NP_001239483.1 | 7 | 1134 | |||
NM_008034.3 | NP_032060.2 | 7 | 1218 | |||
GenBank mRNA | AF096319.1 | - | 7 | 1113 | ||
AK002275.1 | - | 7 | 1213 | |||
BC138781.1 | - | 6 | 851 | 8861803 | ||
BC138795.1 | - | 6 | 938 | 8861817 | ||
BC145414.1 | - | 6 | 938 | 9053972 | ||
M64782.1 | - | 4 | 787 |