Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Folr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252552.1 | NP_001239481.1 | 5 | 672 | ||
NM_001252553.1 | NP_001239482.1 | 5 | 598 | |||
NM_001252554.1 | NP_001239483.1 | 5 | 585 | |||
NM_008034.3 | NP_032060.2 | 5 | 669 | |||
GenBank mRNA | AF096319.1 | - | 5 | 564 | ||
AK002275.1 | - | 5 | 666 | |||
BC002071.1 | - | 2 | 238 | |||
BC023477.1 | - | 2 | 219 | |||
BC037078.1 | - | 4 | 410 | |||
BC138781.1 | - | 4 | 302 | 8861803 | ||
BC138795.1 | - | 4 | 389 | 8861817 | ||
BC145414.1 | - | 4 | 389 | 9053972 | ||
CF617679.1 | - | 4 | 415 | |||
M64782.1 | - | 2 | 238 |