Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fshb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008045.2 | NP_032071.1 | 3 | 278 | ||
GenBank mRNA | AK017593.1 | - | 3 | 279 | ||
BC061159.1 | - | 3 | 278 | 30251083 |