Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G0s2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008059.3 | NP_032085.1 | 2 | 249 | ||
GenBank mRNA | AK003165.1 | - | 2 | 171 | ||
BC117038.1 | - | 2 | 77 | 40126357 | ||
BC117040.1 | - | 2 | 77 | 40126359 | ||
BY124107.1 | - | 2 | 249 | |||
X95280.1 | - | 2 | 153 |