Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008073.3 | NP_032099.1 | 4 | 914 | ||
NM_177408.6 | NP_803127.3 | 4 | 914 | |||
GenBank mRNA | AF233775.1 | - | 4 | 530 | ||
AF233776.1 | - | 4 | 530 | |||
AF233777.1 | - | 4 | 530 | |||
AF233778.1 | - | 4 | 530 | |||
AF233779.1 | - | 4 | 530 | |||
AF233780.1 | - | 4 | 530 | |||
AF233781.1 | - | 4 | 530 | |||
AF233782.1 | - | 4 | 530 | |||
AF233783.1 | - | 4 | 530 | |||
AF233784.1 | - | 4 | 530 | |||
AF233785.1 | - | 4 | 530 | |||
AF233786.1 | - | 4 | 530 | |||
AF233787.1 | - | 4 | 530 | |||
AF233788.1 | - | 4 | 530 | |||
AF233789.1 | - | 4 | 530 | |||
AF233790.1 | - | 4 | 530 | |||
AF233791.1 | - | 4 | 530 | |||
AF233792.1 | - | 4 | 530 | |||
AF233793.1 | - | 4 | 530 | |||
AF233794.1 | - | 4 | 530 | |||
AF233795.1 | - | 4 | 530 | |||
AF233796.1 | - | 4 | 530 | |||
AF233797.1 | - | 4 | 530 | |||
AF233798.1 | - | 4 | 530 | |||
AF233799.1 | - | 4 | 530 | |||
AF233800.1 | - | 4 | 530 | |||
AF233801.1 | - | 4 | 530 | |||
AF233802.1 | - | 4 | 530 | |||
AK034338.1 | - | 4 | 884 | |||
AK047085.1 | - | 4 | 492 | |||
AK049857.1 | - | 4 | 557 | |||
AK136889.1 | - | 4 | 522 | |||
AK141129.1 | - | 4 | 556 | |||
BC031762.1 | - | 4 | 732 | 4504205 | ||
M62374.1 | - | 4 | 877 | |||
M86572.1 | - | 4 | 706 |
Gene Symbol: A230004M16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK049150.1 | - | 6 | 620 |