Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008076.3 | NP_032102.2 | 8, 9 | 1221 | ||
GenBank mRNA | AK132642.1 | - | 8, 9 | 1049 | ||
AK160446.1 | - | 6, 7 | 888 | |||
BC057957.1 | - | 8, 9 | 1206 | 5357713 |