Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144512.2 | NP_653095.1 | 10 | 1166 | ||
GenBank mRNA | AK149557.1 | - | 10 | 1174 | ||
BC021606.1 | - | 1 | 24 | |||
BC023117.1 | - | 10 | 1154 | 4240641 | ||
BC029637.1 | - | 10 | 1166 | 4195373 |