Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK134525.1 | - | 1, 2 | 163 | ||
AK163544.1 | - | 1, 2 | 194 | |||
BB868849.1 | - | 1, 2 | 221 | |||
BY289084.1 | - | 3, 4 | 384 | |||
CX239267.1 | - | 3, 4 | 376 | |||
M96265.1 | - | 3, 4 | 410 |