Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gata3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008091.3 | NP_032117.1 | 3 | 1177 | ||
GenBank mRNA | AK090089.1 | - | 3 | 1186 | ||
AK155549.1 | - | 3 | 1178 | |||
AK156634.1 | - | 3 | 1164 | |||
AK157625.1 | - | 797 | ||||
AK157939.1 | - | 3 | 1051 | |||
AK161099.1 | - | 3 | 1055 | |||
BC062915.1 | - | 2 | 773 | 6826352 | ||
X55123.1 | - | 2 | 793 |