Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gata6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010258.3 | NP_034388.2 | 2 | 1047 | ||
GenBank mRNA | AF179425.1 | - | 1 | 354 | ||
AK053151.1 | - | 2 | 1050 | |||
AK142381.1 | - | 2 | 907 | |||
S82462.1 | - | 1 | 806 |