Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008096.2 | NP_032122.1 | 9 | 1325 | ||
GenBank mRNA | AI876501.1 | - | 5 | 489 | ||
AK010965.1 | - | 9 | 1149 | |||
BC010762.1 | - | 9 | 1188 | 4234788 | ||
BC051395.1 | - | 9 | 1192 | 5125043 | ||
M55413.1 | - | 9 | 1135 |