Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008096.2 | NP_032122.1 | 10 | 1412 | ||
GenBank mRNA | AI876501.1 | - | 4 | 400 | ||
AK010965.1 | - | 10 | 1236 | |||
BC010762.1 | - | 10 | 1275 | 4234788 | ||
BC051395.1 | - | 10 | 1279 | 5125043 | ||
M55413.1 | - | 10 | 1222 |