Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163282.2 | NP_001156754.1 | 7 | 1701 | ||
NM_008107.4 | NP_032133.2 | 1 | 379 | |||
GenBank mRNA | AK162823.1 | - | 7 | 1684 | ||
BC079555.1 | - | 2 | 630 | 30359238 | ||
BC094351.1 | - | 7 | 1653 | |||
M57639.1 | - | 1 | 379 | |||
M62301.1 | - | 7 | 1689 |
Gene Symbol: Cers1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138647.3 | NP_619588.1 | 7 | 1701 | ||
GenBank mRNA | AK053885.1 | - | 7 | 1706 |