Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfra4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136063.2 | NP_001129535.1 | 4 | 620 | ||
NM_001271001.1 | NP_001257930.1 | 4 | 620 | |||
NM_001271002.1 | NP_001257931.1 | 4 | 647 | |||
NM_020014.2 | NP_064398.1 | 4 | 647 | |||
GenBank mRNA | AB041809.1 | - | 2 | 380 | ||
AJ276514.1 | - | 4 | 647 | |||
AJ276516.1 | - | 4 | 647 | |||
AJ276870.1 | - | 4 | 620 | |||
AJ276871.1 | - | 4 | 620 | |||
AK090192.1 | - | 2 | 285 | |||
BC111808.1 | - | 4 | 646 | |||
BC141873.1 | - | 4 | 642 | |||
BC141874.1 | - | 4 | 647 | 40040202 |