Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsc22d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077364.1 | NP_001070832.1 | 4 | 674 | ||
NM_010286.4 | NP_034416.3 | 3 | 499 | |||
GenBank mRNA | AF024519.1 | - | 3 | 484 | ||
AF201287.1 | - | 3 | 243 | |||
AF201288.1 | - | 3 | 479 | |||
AF201289.1 | - | 4 | 636 | |||
AK083389.1 | - | 3 | 498 | |||
AK144196.1 | - | 3 | 502 | |||
BC028813.1 | - | 3 | 489 | 5136440 | ||
BY071907.1 | - | 3 | 316 | |||
EU818782.1 | - | 4 | 673 |