Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gli1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010296.2 | NP_034426.2 | 13 | 3252 | ||
GenBank mRNA | AB025922.1 | - | 13 | 3262 | ||
BC131650.1 | - | 12 | 3230 | 40110429 |