Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010307.3 | NP_034437.1 | 7 | 848 | ||
NM_177137.5 | NP_796111.2 | 7 | 1013 | |||
GenBank mRNA | AK158831.1 | - | 7 | 849 | ||
AY179169.1 | - | 7 | 582 | |||
BC078439.1 | - | 7 | 975 | 5685967 |