Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010312.4 | NP_034442.1 | 7 | 635 | ||
GenBank mRNA | AK146941.1 | - | 7 | 659 | ||
AK151628.1 | - | 7 | 625 | |||
BC029077.1 | - | 6 | 475 | 4019292 | ||
BC059942.1 | - | 7 | 661 | 6529685 | ||
BC062178.1 | - | 7 | 618 | 6330944 | ||
U34960.1 | - | 6 | 455 |