Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013531.4 | NP_038559.2 | 4, 5 | 347 | ||
GenBank mRNA | AF277893.1 | - | 3, 4 | 211 | ||
AF501886.1 | - | 3, 4 | 211 | |||
AK036816.1 | - | 4, 5 | 349 | |||
AK098139.1 | - | 4, 5 | 280 | |||
AK152227.1 | - | 4, 5 | 338 | |||
AK155309.1 | - | 4, 5 | 406 | |||
AK167416.1 | - | 4, 5 | 380 | |||
AK168302.1 | - | 4, 5 | 303 | |||
AK169752.1 | - | 4, 5 | 338 | |||
AK172147.1 | - | 5, 6 | 480 | |||
AK187079.1 | - | 1, 2 | 111 | |||
AK198808.1 | - | 1, 2 | 111 | |||
BC028753.1 | - | 4, 5 | 318 | 1348753 | ||
M63658.1 | - | 3, 4 | 251 | |||
M87286.1 | - | 3, 4 | 251 | |||
S86124.1 | - | 3, 4 | 251 |