Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnrh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008145.2 | NP_032171.1 | 2 | 120 | ||
GenBank mRNA | AK139480.1 | - | 2 | 82 | ||
BC116897.1 | - | 1 | 92 | 40126216 | ||
BC116899.1 | - | 1 | 92 | 40126218 | ||
CJ059442.1 | - | 2 | 82 | |||
CX763652.1 | - | 2 | 120 |