Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnrh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008145.2 | NP_032171.1 | 3, 4 | 338 | ||
GenBank mRNA | AK139480.1 | - | 3, 4 | 300 | ||
BC116897.1 | - | 2, 3 | 310 | 40126216 | ||
BC116899.1 | - | 2, 3 | 310 | 40126218 | ||
CJ059442.1 | - | 3, 4 | 300 | |||
CX763652.1 | - | 3, 4 | 338 |