Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnrhr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010323.1 | NP_034453.1 | 1 | 206 | ||
GenBank mRNA | BC061250.1 | - | 1 | 182 | 30249439 | |
L01119.1 | - | 1 | 206 | |||
L28756.1 | - | 242 | ||||
L28757.1 | - | 1 | 161 | |||
L28758.1 | - | 1 | 161 | |||
M93108.1 | - | 1 | 203 |
Set Membership: |
G-protein coupled receptor |