Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081088.1 | NP_001074557.1 | 17 | 2633 | ||
GenBank mRNA | AK084165.1 | - | 17 | 2597 | ||
AK165434.1 | - | 17 | 2595 |