Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpm6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177955.1 | NP_001171426.1 | 8 | 1401 | ||
NM_001177956.1 | NP_001171427.1 | 8 | 2962 | |||
NM_001177957.1 | NP_001171428.1 | 9 | 1492 | |||
NM_001177958.1 | NP_001171429.1 | 7 | 1281 | |||
NM_001177959.1 | NP_001171430.1 | 7 | 2842 | |||
NM_001177960.1 | NP_001171431.1 | 8 | 1372 | |||
NM_001177961.1 | NP_001171432.1 | 7 | 2507 | |||
NM_001177962.1 | NP_001171433.1 | 8 | 1037 | |||
NM_023122.3 | NP_075611.1 | 7 | 946 | |||
GenBank mRNA | AF254869.1 | - | 7 | 1142 | ||
AF254870.1 | - | 8 | 1262 | |||
AF254871.1 | - | 7 | 917 | |||
AF254872.1 | - | 8 | 1233 | |||
AF254873.1 | - | 7 | 2702 | |||
AF254874.1 | - | 9 | 1353 | |||
AF254875.1 | - | 8 | 2822 | |||
AF254876.1 | - | 8 | 1008 | |||
AF254877.1 | - | 7 | 2477 | |||
AK016567.1 | - | 7 | 871 | |||
AK140714.1 | - | 7 | 961 | |||
S65737.1 | - | 7 | 938 |