Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080743.1 | NP_001074212.1 | 1 | 601 | ||
NM_019497.2 | NP_062370.2 | 1 | 601 | |||
GenBank mRNA | AK078475.1 | - | 1 | 473 | ||
AK132685.1 | - | 1 | 602 | |||
BC150691.1 | - | 1 | 125 | 9087602 | ||
BY719486.1 | - | 1 | 473 |