Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aes | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276288.1 | NP_001263217.1 | 7 | 723 | ||
NM_010347.4 | NP_034477.1 | 7 | 726 | |||
GenBank mRNA | AK019397.1 | - | 1 | 149 | ||
AK131845.1 | - | 7 | 603 | |||
AK149015.1 | - | 4 | 1394 | |||
AK158714.1 | - | 7 | 611 | |||
AK189802.1 | - | 1 | 99 | |||
AK207707.1 | - | 1 | 98 | |||
BC132217.1 | - | 7 | 614 | 40130494 | ||
BC132219.1 | - | 7 | 614 | 40130496 | ||
BC145299.1 | - | 7 | 611 | 9053853 | ||
L12140.1 | - | 7 | 567 | |||
X73359.1 | - | 6 | 677 | |||
X73361.1 | - | 7 | 626 |