Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gss | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291111.1 | NP_001278040.1 | 9 | 992 | ||
NM_008180.2 | NP_032206.1 | 10 | 1127 | |||
GenBank mRNA | AK033435.1 | - | 4 | 1602 | ||
AK146486.1 | - | 10 | 1032 | |||
AK149577.1 | - | 9 | 897 | |||
AK159630.1 | - | 10 | 1052 | |||
AK169096.1 | - | 10 | 1047 | |||
BC003784.1 | - | 10 | 1040 | 3593913 | ||
U35456.1 | - | 9 | 1023 |