Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010358.5 | NP_034488.1 | 5 | 531 | ||
GenBank mRNA | BC003822.1 | - | 5 | 354 | 3600444 | |
BC046758.1 | - | 5 | 384 | 5693552 | ||
BC091763.1 | - | 5 | 364 | 5102093 | ||
J03952.1 | - | 5 | 345 | |||
J04632.1 | - | 5 | 532 |