Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Guca2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008190.1 | NP_032216.1 | 3 | 347 | ||
GenBank mRNA | AK075814.1 | - | 3 | 366 | ||
BC012640.1 | - | 3 | 358 | 4219424 | ||
L05516.1 | - | 3 | 341 | |||
M95175.1 | - | 3 | 347 |