Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Guca2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008191.2 | NP_032217.2 | 1 | 149 | ||
GenBank mRNA | AK002364.1 | - | 1 | 121 | ||
BC024373.1 | - | 1 | 116 | 4971289 | ||
CF423670.1 | - | 1 | 162 | |||
U67800.1 | - | 1 | 105 |