Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Guca2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008191.2 | NP_032217.2 | 2 | 179 | ||
GenBank mRNA | AK002364.1 | - | 2 | 151 | ||
BC024373.1 | - | 2 | 146 | 4971289 | ||
CF423670.1 | - | 2 | 192 | |||
U67800.1 | - | 2 | 135 | |||
U90727.1 | - | 2 | 181 |