Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gzma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010370.2 | NP_034500.1 | 3 | 342 | ||
GenBank mRNA | AK156728.1 | - | 3 | 341 | ||
BC061146.1 | - | 3 | 342 | 30296854 | ||
M13226.1 | - | 4 | 501 | |||
X14799.1 | - | 3 | 344 |