Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H1f0 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008197.3 | NP_032223.2 | 1 | 999 | ||
GenBank mRNA | AK051213.1 | - | 1 | 762 | ||
AK077473.1 | - | 1 | 676 | |||
AK090029.1 | - | 1 | 765 | |||
AK145675.1 | - | 1 | 651 | |||
AK146077.1 | - | 1 | 762 | |||
AK153990.1 | - | 1 | 763 | |||
AK155312.1 | - | 1 | 694 | |||
BC003830.1 | - | 1 | 748 | 3600538 | ||
BC011493.1 | - | 1 | 746 | 4166167 | ||
BC110361.1 | - | 1 | 751 | 5343233 | ||
X13171.1 | - | 1 | 729 |