Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hadh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008212.4 | NP_032238.2 | 4 | 625 | ||
GenBank mRNA | AK075609.1 | - | 4 | 551 | ||
AK132260.1 | - | 4 | 588 | |||
AK148481.1 | - | 3 | 3680 | |||
AK148486.1 | - | 4 | 562 | |||
AK167024.1 | - | 4 | 552 | |||
AK168238.1 | - | 4 | 572 | |||
AK168877.1 | - | 4 | 552 | |||
AK169238.1 | - | 2 | 181 | |||
AK169261.1 | - | 4 | 591 | |||
AK211835.1 | - | 1 | 89 | |||
BC064712.1 | - | 4 | 567 | 30303218 | ||
D29639.1 | - | 4 | 551 |