Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hes3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008237.4 | NP_032263.2 | 3 | 397 | ||
GenBank mRNA | AK090218.1 | - | 3 | 398 | ||
BC116444.2 | - | 2 | 312 | 40104666 |